Learn More
ACROBiosystems ACROBiosystems Human ANGPTL7 / CDT6 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems AN7H5252100UG
Encompass_Preferred
Recombinant Protein;100UG;Human ANGPTL7, Mouse IgG2a Fc Tag (AN7-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Pro 346 (Accession # O43827-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 64.1 kDa. The protein migrates as 60 kDa and 76 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6317