Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ANGPTL8/Betatrophin (NP_061157) VersaClone cDNA

Catalog No. RDC1636
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC1636 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC1636 Supplier R&D Systems™ Supplier No. RDC1636
Add to Cart
Add to Cart
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

The Human ANGPTL8/Betatrophin (NP_061157) VersaClone cDNA clone contains the complete ORF for Angiopoietin-like protein 8/Betatrophin, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Angiopoietin-like protein 8/Betatrophin insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

Specifications

Gene Angiopoietin-like protein 8/Betatrophin
Accession Number NP_061157
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.307
Insert Type 610bp
Gene ID (Entrez) 55908
Gene Alias Angiopoietin like Protein 8, ANGPTL8, Betatrophin, C19orf80, chromosome 19 open reading frame 80, Gm6484, hepatocellular carcinoma-associated gene TD26, hepatocellular carcinoma-associated protein TD26, Lipasin, PRO1185, PVPA599, RIFL, TD26, UNQ599
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg

For Reasearch Use Only. Not for use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.