Learn More
Biomatik Corporation Human Anion/Sugar Transporter (AST) ELISA Kit
Supplier:  Biomatik Corporation EKU02418
Encompass_Preferred
Human Anion/Sugar Transporter (AST) ELISA Kit for detecting AST in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NRA2. Gene ID: 26503. Target Synonyms: SLC17A5; SD; ISSD; NSD; SIALIN; SIASD; SLD; Solute Carrier Family 17 Member 5; Sialic Acid Storage Disease; Vesicular H (+) /Aspartate-glutamate cotransporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9134