Learn More
Biomatik Corporation Human Annexin A11 (ANXA11) ELISA Kit
Supplier:  Biomatik Corporation EKU02426
Encompass_Preferred
Human Annexin A11 (ANXA11) ELISA Kit for detecting ANXA11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50995. Gene ID: 311. Target Synonyms: ANX-A11; ANX11; CAP50; 56 kDa autoantigen; Calcyclin-associated annexin 50. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9132