Learn More
ACROBiosystems ACROBiosystems Human Annexin A5 Protein, His Tag
Catalog No. 502106316
Encompass_Preferred
Recombinant Protein;100UG;Human Annexin A5, His Tag (AN5-H5123) is expressed from E.coli cells. It contains AA Met 1 - Asp 320 (Accession # P08758-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.8 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-6316 Supplier ACROBiosystems Supplier No. AN5H5123100UG
Recombinant Protein;100UG;Human Annexin A5, His Tag (AN5-H5123) is expressed from E.coli cells. It contains AA Met 1 - Asp 320 (Accession # P08758-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.8 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).