Learn More
Biomatik Corporation Human Annexin A8 (ANXA8) ELISA Kit
Supplier:  Biomatik Corporation EKU11763
Encompass_Preferred
Human Annexin A8 (ANXA8) ELISA Kit for detecting ANXA8 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13928. Gene ID: 653145. Target Synonyms: ANX8; ANX-A8; Vascular anticoagulant-beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5641