Learn More
American Research Products Inc Human AnneXin V ANX V ELISA K
Catalog No. 5000329921
Encompass
Human AnneXin V ANX-V ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich
Catalog No. 50-003-29921 Supplier American Research Products Inc Supplier No. CSBE04882H
Human AnneXin V ANX-V ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates cell lysates Method Sandwich