Learn More
Biomatik Corporation Human Anoctamin 6 (ANO6) ELISA Kit
Supplier:  Biomatik Corporation EKU02438
Encompass_Preferred
Human Anoctamin 6 (ANO6) ELISA Kit for detecting ANO6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4KMQ2. Gene ID: 196527. Target Synonyms: TMEM16F; SCAN channel; Transmembrane Protein 16F; Small-conductance calcium-activated nonselective cation channel. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9131