Learn More
Sino Biological Anti-CD13 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10051MM19100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD13 /ANPEP (rh CD13 /ANPEP; Catalog#10051-H08H; NP_001141.2; Lys69-Lys967,603Met/Ile). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2370