Learn More
Sino Biological Anti-CD13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10051T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD13 / ANPEP (rh CD13 / ANPEP; Catalog#10051-H08H; NP_001141.2; Lys69-Lys967,603Met/Ile). CD13 / ANPEP specific IgG was purified by Human CD13 / ANPEP affinity chromatography.
Catalog No. 50-240-9073