Learn More
Biomatik Corporation Human Anthrax Toxin Receptor 1 (ANTXR1) ELISA Kit
Supplier:  Biomatik Corporation EKU11992
Encompass_Preferred
Human Anthrax Toxin Receptor 1 (ANTXR1) ELISA Kit for detecting ANTXR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9H6X2. Gene ID: 84168. Target Synonyms: ATR; TEM8; Tumor Endothelial Marker 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8054