Learn More
Biomatik Corporation Human Anthrax Toxin Receptor 2 (ANTXR2) ELISA Kit
Supplier:  Biomatik Corporation EKU02440
Encompass_Preferred
Human Anthrax Toxin Receptor 2 (ANTXR2) ELISA Kit for detecting ANTXR2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58335. Gene ID: 118429. Target Synonyms: CMG-2; CMG2; ISH; JHF; Capillary Morphogenesis Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4525