Learn More
Abcam Human Anti-Ascaris lumbricoides IgG ELISA Kit, 1 x 96 Tests
Catalog No. 502658497
Encompass_Preferred
Human Anti-Ascaris lumbricoides IgG ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Ascaris lumbricoides IgG in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-8497 Supplier Abcam Supplier No. AB1087071X96TESTS
Human Anti-Ascaris lumbricoides IgG ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Ascaris lumbricoides IgG in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement