Learn More
Abcam Human Anti-Brucella IgM ELISA Kit, 1 x 96 Tests
Catalog No. 502653501
Encompass_Preferred
Human Anti-Brucella IgM ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Brucella IgM in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-3501 Supplier Abcam Supplier No. AB1087131X96TESTS
Human Anti-Brucella IgM ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Brucella IgM in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement