Learn More
Abcam Human Anti-Cytomegalovirus IgG ELISA Kit (CMV), 1 x 96 Tests
Supplier:  Abcam AB1087241X96TESTS
Encompass_Preferred
Anti-cytomegalovirus (CMV) IgG ELISA kit is designed for the qualitative detection of IgG antibodies against cytomegalovirus in human samples - Colorimetric assay - 450 nm readout - works on any plate reader - validated in serum and plasma - 98% sensitivity (probability of the assay scoring positive in the presence of the specific analyte) - Cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1584333