Learn More
Abcam Human Anti-Epstein Barr virus IgG ELISA Kit (EBV-EBNA), 1 x 96 Tests
Supplier:  Abcam AB1087311X96TESTS
Encompass_Preferred
Human Anti-Epstein Barr virus IgG ELISA Kit (EBV-EBNA) is a Indirect ELISA kit for the measurement of Human Anti-Epstein Barr virus IgG (EBV-EBNA) in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1850831