Learn More
Abcam Human Anti-Herpes simplex virus Type 1 and 2 IgM ELISA Kit (HSV1+2), 1 x 96 Tests
Catalog No. 502653498
Encompass_Preferred
Human Anti-Herpes simplex virus Type 1 and 2 IgM ELISA Kit (HSV1+2) is a Indirect ELISA kit for the measurement of Human Anti-Herpes simplex virus Type 1 and 2 IgM (HSV1+2) in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-3498 Supplier Abcam Supplier No. AB1087421X96TESTS
Human Anti-Herpes simplex virus Type 1 and 2 IgM ELISA Kit (HSV1+2) is a Indirect ELISA kit for the measurement of Human Anti-Herpes simplex virus Type 1 and 2 IgM (HSV1+2) in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement