Learn More
ACROBiosystems Human Anti-Human IgM 10mg
Catalog No. 502534412
Encompass_Preferred
The Anti-Human IgM-coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal anti-human IgM antibody. The beads can be used to capture the Human IgM in Chemiluminescence procedures. It has no-corss-reactivity with Human IgG IgA or IgE. The Anti-Human IgM-coupled Magnetic Beads is easy to capture the human IgM and the bounded antibody have no activity lost this ready to use products could greatly save your protein coupling time and hassle and help us get the best performance and highly reproducible results.
Catalog No. 50-253-4412 Supplier ACROBiosystems Supplier No. MPCA01010MG
The Anti-Human IgM-coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal anti-human IgM antibody. The beads can be used to capture the Human IgM in Chemiluminescence procedures. It has no-corss-reactivity with Human IgG IgA or IgE. The Anti-Human IgM-coupled Magnetic Beads is easy to capture the human IgM and the bounded antibody have no activity lost this ready to use products could greatly save your protein coupling time and hassle and help us get the best performance and highly reproducible results.