Learn More
Abcam Human Anti-Parainfluenza virus 1,2,3 IgA ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1087571X96TESTS
Encompass_Preferred
Human Anti-Parainfluenza virus 1,2,3 IgA ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Parainfluenza virus 1,2,3 IgA in Human in Citrate plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2611373