Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals, LLC Human anti-SARS-CoV-2 IgG ELISA Kit

Catalog No. MP08440100 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08440100 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08440100 Supplier MP Biomedicals, LLC Supplier No. 08440100
Add to Cart
Add to Cart

In vitro quantitative determination of anti-SARS-CoV-2 IgG concentrations from human samples of plasma, tissue homogenates, and other biological fluids.

  • Allows for the in vitro quantitative determination of anti-SARS-CoV-2 IgG concentrations
  • Sample Type: Human plasma, tissue homogenates, and other biological fluids
  • Range: 3.9-250 ng/mL
  • Sensitivity: 2.34 ng/mL
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.9 to 250 ng/mL
Assay Sensitivity 2.34 ng/mL
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA Assays
Sample Type Biological Fluids
Target Species Human
For Use With (Equipment) Microplate Reader, Automated Plate Washer
Immunoassay Kit Grade Ultrasensitive
Label or Dye HRP
Quantity 96 Tests
Target Anti-SARS-CoV-2 IgG
Target Organism Class Protein
Target Species Validated Human
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.