Learn More
Biomatik Corporation Human Apolipoprotein (Anti-Apo) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU02442
Encompass_Preferred
Human Apolipoprotein (Anti-Apo) Anti-antigen ELISA Kit for detecting Anti-Apo in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.29ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P02647. Gene ID: 335. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4874