Learn More
Biomatik Corporation Human Cardiac Troponin I (cTnI) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12134
Encompass_Preferred
Human Cardiac Troponin I (cTnI) Anti-antigen ELISA Kit for detecting cTnI in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P19429. Gene ID: 7137. Target Synonyms: TNNI3; TNNC1; CMH7; C-TnI; Troponin I Type 3; Troponin I Type 3, Cardiac. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5333