Learn More
Biomatik Corporation Human Chloride Intracellular Channel Protein 1 (CLIC1) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11553
Encompass_Preferred
Human Chloride Intracellular Channel Protein 1 (CLIC1) Anti-antigen ELISA Kit for detecting CLIC1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: O00299. Gene ID: 1192. Target Synonyms: G6; NCC27; p64CLCP; hRNCC; Chloride channel ABP; Nuclear chloride ion channel 27; Regulatory nuclear chloride ion channel protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5804