Learn More
Biomatik Corporation Human Coagulation Factor II (F2) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU11551
Encompass_Preferred
Human Coagulation Factor II (F2) Anti-antigen ELISA Kit for detecting F2 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P00734. Gene ID: 2147. Target Synonyms: FII; TM; PT; Thrombin; Prothrombin; Pro-Thrombin; Activation peptide fragment 1; Activation peptide fragment 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8125