Learn More
Biomatik Corporation Human Epidermal Growth Factor Receptor (EGFR) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12148
Encompass_Preferred
Human Epidermal Growth Factor Receptor (EGFR) Anti-antigen ELISA Kit for detecting EGFR in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P00533. Gene ID: 1956. Target Synonyms: ErbB-1; ErbB1; ER1; ERB-B1; MENA; HER1; Erythroblastic Leukemia Viral (v-Erb-B) oncogene Homolog,Avian; Proto-oncogene c-ErbB-1; Receptor tyrosine-protein kinase erbB-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5227