Learn More
Biomatik Corporation Human Folate Receptor 1, Adult (FOLR1) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12456
Encompass_Preferred
Human Folate Receptor 1, Adult (FOLR1) Anti-antigen ELISA Kit for detecting FOLR1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.19ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P15328. Gene ID: 2348. Target Synonyms: FBP; FOLR; FR-alpha; MOv18; Adult folate-binding protein; KB cells FBP; Ovarian tumor-associated antigen MOv18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6302