Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B (GRIN2B) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12521
Encompass_Preferred
Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2B (GRIN2B) Anti-antigen ELISA Kit for detecting GRIN2B in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.23ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: Q13224. Gene ID: 2904. Target Synonyms: NMDAR2B; NR2B; hNR3; Glutamate [NMDA] receptor subunit epsilon-2; N-methyl D-aspartate receptor subtype 2B; N-methyl-D-aspartate receptor subunit 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6042