Learn More
Biomatik Corporation Human Glycyl tRNA Synthetase (GARS) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12465
Encompass_Preferred
Human Glycyl tRNA Synthetase (GARS) Anti-antigen ELISA Kit for detecting GARS in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.33ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P41250. Gene ID: 2617. Target Synonyms: CMT2D; CMT2-D; DSMAV; GlyRS; SMAD1; EJ; Charcot-Marie-Tooth Neuropathy 2D; Glycine tRNA Ligase; Diadenosine tetraphosphate synthetase; AP-4-A synthetase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5779