Learn More
Biomatik Corporation Human Interleukin 1 Beta (IL1b) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12139
Encompass_Preferred
Human Interleukin 1 Beta (IL1b) Anti-antigen ELISA Kit for detecting IL1b in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.13ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P01584. Gene ID: 3553. Target Synonyms: IL1-B; IL1-Beta; IL1F2; IL-1β; Interleukin-1 Family Member 2; Catabolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7206