Learn More
Biomatik Corporation Human Mesothelin (MSLN) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12066
Encompass_Preferred
Human Mesothelin (MSLN) Anti-antigen ELISA Kit for detecting MSLN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: Q13421. Gene ID: 10232. Target Synonyms: SMR; MPF; CAK1; Pre-pro-megakaryocyte-potentiating factor; Megakaryocyte-potentiating factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8042