Learn More
Biomatik Corporation Human Osteopontin (OPN) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12161
Encompass_Preferred
Human Osteopontin (OPN) Anti-antigen ELISA Kit for detecting OPN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P10451. Gene ID: 6696. Target Synonyms: SPP1; BNSP; BSPI; ETA1; Secreted Phosphoprotein 1; Bone Sialoprotein I; Early T-Lymphocyte Activation 1; Nephropontin; Urinary stone protein; Uropontin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4772