Learn More
Biomatik Corporation Human Pentraxin 3, Long (PTX3) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU10645
Encompass_Preferred
Human Pentraxin 3, Long (PTX3) Anti-antigen ELISA Kit for detecting PTX3 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P26022. Gene ID: 5806. Target Synonyms: TNFAIP5; TSG-14; Pentaxin-Related Gene,Rapidly Induced By IL-1 Beta; Tumor Necrosis Factor Alpha-Induced Protein 5; Tumor necrosis factor-inducible gene 14 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5259