Learn More
Biomatik Corporation Human Polymeric Immunoglobulin Receptor (PIGR) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12515
Encompass_Preferred
Human Polymeric Immunoglobulin Receptor (PIGR) Anti-antigen ELISA Kit for detecting PIGR in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.18ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P01833. Gene ID: 5284. Target Synonyms: SC; Secretory Component; Hepatocellular carcinoma-associated protein TB6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5453