Learn More
Biomatik Corporation Human Selenium Binding Protein 1 (SELENBP1) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12055
Encompass_Preferred
Human Selenium Binding Protein 1 (SELENBP1) Anti-antigen ELISA Kit for detecting SELENBP1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.18ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: Q13228. Gene ID: 8991. Target Synonyms: LPSB; SP56; hSBP; hSP56; SBP56; 56 kDa selenium-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8045