Learn More
Enzyme Research Laboratories Inc Human Antithrombin, 1 mg
Supplier:  Enzyme Research Laboratories Inc HAT
Encompass
Purified from fresh frozen human plasma. AT is a single-chain glycoprotein which is considered to be the main physiological inhibitor of thrombin and Factor Xa. In the presence of heparin, the inhibition of thrombin and factor Xa is enhanced from 300 to 1,000 fold. AT purity is >95% pure.
Catalog No. NC0057425