Learn More
Haematologic Technologies Inc HUMAN ANTITHROMBIN III 1MG
Catalog No. 50883407
Encompass_Preferred
Coagulation protein, Human Antithrombin III, 1 milligram, Inhibitor, Mode of action: Serine protease inhibitor, Research use only, Supplied in 50% (v/v) glycerol/water, store at -20 Celsius
Catalog No. 50-883-407 Supplier Haematologic Technologies Inc Supplier No. HCATIII0120
Coagulation protein, Human Antithrombin III, 1 milligram, Inhibitor, Mode of action: Serine protease inhibitor, Research use only, Supplied in 50% (v/v) glycerol/water, store at -20 Celsius