Learn More
Abcam Human Antithrombin III ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1088011X96TESTS
Encompass_Preferred
Human Antithrombin III ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Antithrombin III in Human in Cerebral Spinal Fluid, Milk, Plasma, Saliva, Serum, Urine samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2404862