Learn More
Aniara Diagnostica LLC HUMAN ANTITHROMIN 10X1.5MG
Catalog No. 501009015
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Highly purified human Antithrombin III (MW=58,000 Da) from human plasma, lyophilized with glycine, HEPES and sodium chloride. Specific activity > 6 IU/mg. For in vitro use only. 10 vials x 1.5 mg.
Catalog No. 50-100-9015 Supplier Aniara Diagnostica LLC Supplier No. APP004L
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Highly purified human Antithrombin III (MW=58,000 Da) from human plasma, lyophilized with glycine, HEPES and sodium chloride. Specific activity > 6 IU/mg. For in vitro use only. 10 vials x 1.5 mg.