Learn More
Aniara Diagnostica LLC HUMAN ANTITHROMIN 3.75MG
Catalog No. 501009013
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Highly purified human Antithrombin III (MW=58,000 Da) from human plasma, lyophilized with glycine, HEPES and sodium chloride. Specific activity > 6 IU/mg. For in vitro use only. 1 vial x 3.75 mg.
Catalog No. 50-100-9013 Supplier Aniara Diagnostica LLC Supplier No. APP004D
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Highly purified human Antithrombin III (MW=58,000 Da) from human plasma, lyophilized with glycine, HEPES and sodium chloride. Specific activity > 6 IU/mg. For in vitro use only. 1 vial x 3.75 mg.