Learn More
Biomatik Corporation Human Vinculin (VCL) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12495
Encompass_Preferred
Human Vinculin (VCL) Anti-antigen ELISA Kit for detecting VCL in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P18206. Gene ID: 7414. Target Synonyms: MVCL; Metavinculin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5863