Learn More
Biomatik Corporation Human Vitamin D Binding Protein (DBP) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU12299
Encompass_Preferred
Human Vitamin D Binding Protein (DBP) Anti-antigen ELISA Kit for detecting DBP in serum. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.23ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P02774. Gene ID: 2638. Target Synonyms: GC; VDBG; VDBP; Group-Specific Component; Gc-globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6481