Learn More
Sino Biological Recombinant Human Annexin V/ANXA5 Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 10448HNAEB20
Encompass_Preferred
A DNA sequence encoding the human ANXA5 (P08758) (Met1-Asp320) was expressed and purified. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5298