Learn More
Sino Biological Anti-ANXA9/annexin A9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107589T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ANXA9/annexin A9, and purified by antigen affinity chromatography.
Catalog No. 50-240-8147