Learn More
Sino Biological Anti-AP2B1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106981T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human AP2B1, and purified by antigen affinity chromatography.
Catalog No. 50-240-7859