Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AP3B2 (aa 658-723) Control Fragment Recombinant Protein

Catalog No. RP98025
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98025 Supplier Invitrogen™ Supplier No. RP98025
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111115. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The AP3 complex is a heterotetramer composed of two large adaptins (AP3D1, AP3B1 or AP3B2), a medium adaptin (AP3M1 or AP3M2) and a small adaptin (APS1 or AP3S2). The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes. Unlike the homologous AP3B1, AP3B2-containing AP3 complexes are preferentially targeted to neuronal processes, and mice deficient in AP3B2 displayed compromised synaptic zinc stores.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13367
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 8120
Name Human AP3B2 (aa 658-723) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias [b]-NAP; adapter-related protein complex 3 subunit beta-2; adaptor protein complex AP-3 beta-2 subunit; adaptor protein complex AP-3 subunit beta-2; adaptor related protein complex 3 beta 2 subunit; adaptor related protein complex 3, beta 2 subunit; adaptor-related protein complex 3 subunit beta-2; adaptor-related protein complex 3, beta 2 subunit; adaptor-related protein complex AP-3, beta 2 subunit; AI549966; ap3; AP-3 complex beta-2 subunit; AP-3 complex subunit beta-2; Ap3b2; AU042881; beta3B; beta-3B-adaptin; beta3B-adaptin; beta-NAP; Clathrin a; clathrin assembly protein complex 3 beta-2 large chain; NAPTB; Neuronal adaptin-like protein, beta-subunit; neuron-specific vesicle coat protein beta-NAP
Common Name AP3B2
Gene Symbol AP3B2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VPEWTKCSNREKRKEKEKPFYSDSEGESGPTESADSDPESESESDSKSSSESGSGESSSESDNEDQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.