Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AP3S1 Synthetic Peptide

Catalog No. PEP1275
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1275 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1275 Supplier Invitrogen™ Supplier No. PEP1275
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 19 amino acids near the center of human AP3S1. PEP-1275 can be used as a blocking peptide with polyclonal antibody PA5-21161.

Part of the AP-3 complex, an adapter-related complex which is not clathrin-associated. The complex is associated with the Golgi region as well as more peripheral structures. It facilitates the budding of vesicles from the Golgi membrane and may be directly involved in trafficking to lysosomes.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 1176
Species Human
Purification Method purified
Accession Number Q92572
Common Name AP3S1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias adapter-related protein complex 3 subunit sigma-1; adaptor related protein complex 3 sigma 1 subunit; adaptor related protein complex 3, sigma 1 subunit; adaptor-related protein complex 3 subunit sigma-1; adaptor-related protein complex 3, sigma 1 subunit; AP-3 complex subunit sigma-1; AP-3 complex subunit sigma-3A; AP3S1; CLAPS3; clathrin adaptor complex AP3, sigma-3A subunit; clathrin-associated/assembly/adapter protein, small 3; Clathrin-associated/assembly/adaptor protein, small 3; clathrin-associated/assembly/adaptor protein, small 3 (22kD); Sigma3A; Sigma-3A-adaptin; Sigma3A-adaptin; Sigma-adaptin 3a
Gene Symbol AP3S1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.