Learn More
Sino Biological Anti-FE65/APBB1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107596T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FE65/APBB1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8144