Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human APC10 Synthetic Peptide

Catalog No. PEP1064 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP1064 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP1064 Supplier Invitrogen™ Supplier No. PEP1064
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 16 amino acids near the center of human APC10 PEP-1064 can be used as a blocking peptide with polyclonal antibody PA5-20950.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC10 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. It has been suggested that APC10 plays a role to regulate the binding of specific substrates to the APC/C complex, similar to that of coactivators.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 10393
Species Human
Purification Method purified
Accession Number Q9UM13
Common Name APC10
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 1500026N15Rik; A830003M23Rik; ANAPC10; anaphase promoting complex subunit 10; anaphase-promoting complex subunit 10; APC10; cyclosome subunit 10; DKFZp564L0562; DOC1
Gene Symbol ANAPC10
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.