Learn More
Sino Biological Human APCS Protein 50ug (Sino Biological)
Supplier:  Sino Biological 81346R02H50
Encompass_Preferred
A DNA sequence encoding the Human APCS (P02743) (Met1-Val223) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4541