Learn More
Sino Biological Anti-APEH/AARE Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105219T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human APEH/AARE, and purified by antigen affinity chromatography.
Catalog No. 50-241-2473